missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ EC 23

Catalog No. 401110
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
401110 10 mg
401150 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 401110 Supplier Tocris Bioscience™ Supplier No. 4011/10

Synthetic retinoid; induces differentiation of stem cells

Photostable synthetic retinoid; analog of ATRA (Cat. No. 0695). Displays similar activity to ATRA when tested on the embryonal carcinoma stem cell model TERA2.cl.SP12. Potently induces neural differentiation in human pluripotent embryonic stem cells.

Chemical Identifiers

CAS 104561-41-3
Molecular Formula C23H24O2

Specifications

Quantity 10 mg
Formula Weight 332.44
Purity >99%
Molecular Weight (g/mol) 332.443
Chemical Name or Material EC 23
Recommended Storage Store at +4°C