missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Ebselen | |
| 183°C | |
| C13H9NOSe | |
| 100 mg | |
| ebselen, 2-phenyl-1,2-benzisoselenazol-3 2h-one, ebselene, ebseleno, ebselenum, ebselene french, ebselenum latin, ebselen inn, ebseleno spanish, harmokisane | |
| C1=CC=C(C=C1)N2C(=O)C3=CC=CC=C3[Se]2 | |
| 274.192 | |
| CHEBI:77543 | |
| ≥98.0% (GC) |
| 60940-34-3 | |
| Yellow | |
| MFCD00210937 | |
| 3077 | |
| DYEFUKCXAQOFHX-UHFFFAOYSA-N | |
| 2-phenyl-1,2-benzoselenazol-3-one | |
| 3194 | |
| 274.18 | |
| Crystalline Powder |
Chemical Identifiers
| 60940-34-3 | |
| 274.192 | |
| DYEFUKCXAQOFHX-UHFFFAOYSA-N | |
| 3194 | |
| 2-phenyl-1,2-benzoselenazol-3-one |
| C13H9NOSe | |
| MFCD00210937 | |
| ebselen, 2-phenyl-1,2-benzisoselenazol-3 2h-one, ebselene, ebseleno, ebselenum, ebselene french, ebselenum latin, ebselen inn, ebseleno spanish, harmokisane | |
| CHEBI:77543 | |
| C1=CC=C(C=C1)N2C(=O)C3=CC=CC=C3[Se]2 |
Safety and Handling
RTECSNumber : DE4140750
TSCA : No
Recommended Storage : Refrigerator