missing translation for 'onlineSavingsMsg'
Learn More

Ebselen 98.0+%, TCI America™

Catalog No. p-7129240
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E094625MG 25 mg
E0946100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E094625MG Supplier TCI America Supplier No. E094625MG
Only null left

Chemical Identifiers

CAS 60940-34-3
Molecular Formula C13H9NOSe
Molecular Weight (g/mol) 274.192
MDL Number MFCD00210937
InChI Key DYEFUKCXAQOFHX-UHFFFAOYSA-N
Synonym ebselen, 2-phenyl-1,2-benzisoselenazol-3 2h-one, ebselene, ebseleno, ebselenum, ebselene french, ebselenum latin, ebselen inn, ebseleno spanish, harmokisane
PubChem CID 3194
ChEBI CHEBI:77543
IUPAC Name 2-phenyl-1,2-benzoselenazol-3-one
SMILES C1=CC=C(C=C1)N2C(=O)C3=CC=CC=C3[Se]2

Specifications

Melting Point 183°C
Color Yellow
Quantity 25 mg
UN Number 3077
Formula Weight 274.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ebselen
RTECSNumber DE4140750
TSCA No
Recommended Storage Refrigerator