accessibility menu, dialog, popup

UserName

CAS RN 60940-34-3

IUPAC Name: 2-phenyl-2,3-dihydro-1,2-benzoselenazol-3-one
Synonyms: ebselen
Molecular Weight (g/mol): 274.19
Molecular Formula: C13H9NOSe
InChi Key: DYEFUKCXAQOFHX-UHFFFAOYSA-N
SMILES: O=C1N([Se]C2=CC=CC=C12)C1=CC=CC=C1
Physical Form 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)