missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Streptavidin eFluor™ 660 Conjugate

The streptavidin fluorochrome conjugates are commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry, immunohistochemistry, and immunocytochemistry applications.

$249.00 - $618.00

Specifications

Concentration 0.2 mg/mL
Conjugate eFluor 660
Excitation/Emission 633-647/668 nm
Flow Cytometer Laser Lines Red
Form Liquid
View More Specs

Alert:

Contains Sodium azide


Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124352
View Documents
Invitrogen™
50431780
25 μg
Each for $249.00
Only null left
 
501124353
View Documents
Invitrogen™
50431782
100 μg
Each for $618.00
Only null left
 
Description

Description

The streptavidin fluorochrome conjugates are commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry, immunohistochemistry, and immunocytochemistry applications. Streptavidin binds to biotin with high affinity.

Reported Applications

  • Flow Cytometric Analysis
  • Immunohistochemical Staining of Formalin-Fixed Paraffin Embedded Tissue Sections
  • Immunohistochemical Staining of Frozen Tissue Sections
  • Immunocytochemistry
Specifications

Specifications

0.2 mg/mL
633-647/668 nm
Liquid
Streptavidin Conjugate (fluorescent)
eFluor 660
Red
eBioscience
Biotin
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only.