Learn More
Invitrogen™ eBioscience™ Streptavidin eFluor™ 570 Conjugate
The streptavidin fluorochrome conjugates are commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry, immunohistochemistry, and immunocytochemistry applications.
$227.00 - $512.00
Specifications
| Concentration | 0.2 mg/mL |
|---|---|
| Conjugate | eFluor 570 |
| Excitation/Emission | 555/570 nm |
| Form | Liquid |
| Product Line | eBioscience |
Description
The streptavidin fluorochrome conjugates are commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry, immunohistochemistry, and immunocytochemistry applications. Streptavidin binds to biotin with high affinity.
Reported Applications
- Immunohistochemical Staining of Formalin-Fixed Paraffin Embedded Tissue Sections
- Immunohistochemical Staining of Frozen Tissue Sections
- Immunocytochemistry
Specifications
| 0.2 mg/mL | |
| 555/570 nm | |
| eBioscience | |
| Biotin |
| eFluor 570 | |
| Liquid | |
| Streptavidin Conjugate (fluorescent) |
For Research Use Only. Not for use in diagnostic procedures.