missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Streptavidin eFluor™ 570 Conjugate

Catalog No. 501123611
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501123611 25 μg
501123612 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 501123611 Supplier Invitrogen™ Supplier No. 41431780

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The streptavidin fluorochrome conjugates are commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry, immunohistochemistry, and immunocytochemistry applications.

The streptavidin fluorochrome conjugates are commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry, immunohistochemistry, and immunocytochemistry applications. Streptavidin binds to biotin with high affinity.

Reported Applications

  • Immunohistochemical Staining of Formalin-Fixed Paraffin Embedded Tissue Sections
  • Immunohistochemical Staining of Frozen Tissue Sections
  • Immunocytochemistry

Specifications

Concentration 0.2 mg/mL
Conjugate eFluor 570
Quantity 25 μg
Storage Buffer Aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Synonym SA, Sav
Content And Storage 2°C to 8°C. Do not freeze. Light-sensitive material.
Excitation/Emission 555/570 nm
Form Liquid
Product Line eBioscience
For Use With (Application) Immunohistochemical Staining of Formalin-Fixed Paraffin Embedded Tissue Sections, Immunohistochemical Staining of Frozen Tissue Sections, Immunocytochemistry
Product Type Streptavidin Conjugate (fluorescent)
Target Molecule Biotin
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.