missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Eaton's Reagent, P{2}O{5} ca 7.5% w/w

CAS: 39394-84-8 | CH4O8P2S | 238.043 g/mol

$104.51 - $358.36

Chemical Identifiers

CAS 39394-84-8
Molecular Formula CH4O8P2S
Molecular Weight (g/mol) 238.043
MDL Number MFCD00070545
InChI Key JHNLZOVBAQWGQU-UHFFFAOYSA-N
Synonym eaton's reagent, phosphorus pentoxide methanesulfonic acid, diphosphooxidane; methanesulfonic acid, eaton's, eaton'sreagent, eaton inverted exclamation markas reagent, eaton's reagent, p2o5 ca w/w, phosphorus pentoxide, 7.7 wt. % in methanesulfonic acid
PubChem CID 16149098
SMILES CS(=O)(=O)O.O=P(=O)OP(=O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL15607AE
View Documents
Thermo Scientific Chemicals
L15607AE
100 mL
Each for $104.51
Only null left
 
AAL15607AP
View Documents
Thermo Scientific Chemicals
L15607AP
500 mL
Each for $358.36
Only null left
 
Description

Description

Eaton's Reagent is used in the synthesis fluorine-based hole-transporting material, mono and bis-chalcone derivatives via Claisen-Schmidt condensation reaction between arylaldehydes and ketones and 4-hydroxycoumarins and 4-hydroxy-2-quinolinones derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

39394-84-8
238.043
JHNLZOVBAQWGQU-UHFFFAOYSA-N
16149098
CH4O8P2S
MFCD00070545
eaton's reagent, phosphorus pentoxide methanesulfonic acid, diphosphooxidane; methanesulfonic acid, eaton's, eaton'sreagent, eaton inverted exclamation markas reagent, eaton's reagent, p2o5 ca w/w, phosphorus pentoxide, 7.7 wt. % in methanesulfonic acid
CS(=O)(=O)O.O=P(=O)OP(=O)=O

Specifications

39394-84-8
P2O5
UN3265
eaton's reagent, phosphorus pentoxide methanesulfonic acid, diphosphooxidane; methanesulfonic acid, eaton's, eaton'sreagent, eaton inverted exclamation markas reagent, eaton's reagent, p2o5 ca w/w, phosphorus pentoxide, 7.7 wt. % in methanesulfonic acid
JHNLZOVBAQWGQU-UHFFFAOYSA-N
238.043
141.95
100 mL
Moisture sensitive; May form a precipitate
≈7.5% w/w
CH4O8P2S
MFCD00070545
1.515
Decomposes in water.
CS(=O)(=O)O.O=P(=O)OP(=O)=O
16149098
1.435
>110°C (230°F)
Eaton's Reagent, P2O5
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

P234-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c

H290-H302+H312-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures; Store under Argon

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only