missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Eaton's Reagent, P{2}O{5} ca 7.5% w/w
Description
Eaton's Reagent is used in the synthesis fluorine-based hole-transporting material, mono and bis-chalcone derivatives via Claisen-Schmidt condensation reaction between arylaldehydes and ketones and 4-hydroxycoumarins and 4-hydroxy-2-quinolinones derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Linear Formula | P2O5 |
| UN Number | UN3265 |
| Density | 1.515 |
| Solubility Information | Decomposes in water. |
| Formula Weight | 141.95 |
| Refractive Index | 1.435 |
| Quantity | 500 mL |
| Flash Point | >110°C (230°F) |
| Sensitivity | Moisture sensitive; May form a precipitate |
| Assay | ≈7.5% w/w |
| Show More |
Safety and Handling
| Hazard Category | H290-H302+H312-H314-H335 |
| Hazard Statement | GHS H Statement H314-H318-H302 Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. |
| Precautionary Statement | P234-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S. |
| TSCA | No |
| Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only