missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Eaton's Reagent, P{2}O{5} ca 7.5% w/w

Catalog No. AAL15607AE
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL15607AE 100 mL
AAL15607AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL15607AE Supplier Thermo Scientific Chemicals Supplier No. L15607AE
Only null left

CAS: 39394-84-8 | CH4O8P2S | 238.043 g/mol

Eaton's Reagent is used in the synthesis fluorine-based hole-transporting material, mono and bis-chalcone derivatives via Claisen-Schmidt condensation reaction between arylaldehydes and ketones and 4-hydroxycoumarins and 4-hydroxy-2-quinolinones derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 39394-84-8
Molecular Formula CH4O8P2S
Molecular Weight (g/mol) 238.043
MDL Number MFCD00070545
InChI Key JHNLZOVBAQWGQU-UHFFFAOYSA-N
Synonym eaton's reagent, phosphorus pentoxide methanesulfonic acid, diphosphooxidane; methanesulfonic acid, eaton's, eaton'sreagent, eaton inverted exclamation markas reagent, eaton's reagent, p2o5 ca w/w, phosphorus pentoxide, 7.7 wt. % in methanesulfonic acid
PubChem CID 16149098
SMILES CS(=O)(=O)O.O=P(=O)OP(=O)=O

Specifications

Linear Formula P2O5
UN Number UN3265
Density 1.515
Solubility Information Decomposes in water.
Formula Weight 141.95
Refractive Index 1.435
Quantity 100 mL
Flash Point >110°C (230°F)
Sensitivity Moisture sensitive; May form a precipitate
Assay ≈7.5% w/w
Chemical Name or Material Eaton's Reagent, P2O5
Show More Show Less
Hazard Category H290-H302+H312-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P234-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only