missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EasyPep™ MS Sample Prep Kits

Catalog No. FERA57865
Change view
Click to view available options
Description:
Micro MS Sample Prep Kit
Mini MS Sample Prep Kit
Peptide Clean-up Plate
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
FERA57865 Peptide Clean-up Plate
PIA40006 Mini MS Sample Prep Kit
FERA57864 Micro MS Sample Prep Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. FERA57865 Supplier Thermo Scientific™ Supplier No. A57865
Only null left

Generate high quality proteomic samples for MS analysis in less than four hours using optimized EasyPep MS Sample Prep kits.

EasyPep Mini MS Sample Prep Kits enable efficient and reproducible processing of cultured mammalian cells and tissues for proteomic MS analysis. The kit contains pre-formulated buffers, MS-grade enzyme mix, peptide clean-up columns, and an optimized protocol to generate MS-compatible peptide samples in less than three hours. Kits are optimized to process protein samples from 10–100 μg with a high yield of MS-ready peptides.

Complete—includes pre-formulated reagents for lysis through digestion, peptide clean-up columns, and an optimized protocol for up to 20 samples
Optimized—streamlined protocol and reagents minimize the number of steps and time time needed to process samples
Flexible—reagents and protocol have been verified using cells, plasma, and tissue samples for 10 to 100 μg samples
Time-saving—sample processing time reduced from more than one day to less than three hours
Compatible—final preparation is ready for direct MS analysis and other downstream applications, including TMT labeling

The sample preparation of peptides for MS analysis is complex, with numerous steps and varied protocols, resulting in sample variability in both quality and quantity. This kit has been designed to improve reproducibility while saving hands-on and processing time. The number of steps and processing time have been reduced through the addition of Universal Nuclease to reduce viscosity from nucleic acids (replacing sonication), a rapid 'one pot' reduction/alkylation solution for cysteine modification (carbamidomethylation, + 57.02), and a trypsin/Lys-C protease mix for more complete digestion. In addition, the kit includes Peptide Clean-up columns and buffers to prepare detergent-free peptide samples for MS analysis. The optimized reagents and protocol produce high-quality peptides that are compatible with TMT labeling and other downstream applications, or are ready for MS analysis.

Specifications

Description Peptide Clean-up Plate
Product Type Reagent Kit
Product Line EasyPep
Starting Material Protein Samples
Detection Method Mass Spectrometry
Format Kit
Workflow Step Mass spectrometry sample preparation
Quantity 96 reactions
Form Solid