missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EasyPep™ Magnetic MS Sample Preparation Kits and Proteomics Magnetic Clean-up Beads

Catalog No. FERA57866
Change view
Click to view available options
Quantity:
20 Reactions
96 Reactions
5 mL
Product Type:
Magnetic Clean-up Beads
Magnetic MS Sample Prep Kit
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
FERA57866 20 Reactions Magnetic MS Sample Prep Kit
FERA57867 96 Reactions Magnetic MS Sample Prep Kit
FERA57868 5 mL Magnetic Clean-up Beads
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. FERA57866 Supplier Thermo Scientific™ Supplier No. A57866

EasyPep Magnetic MS Sample Preparation Kits and Proteomics Magnetic Clean-up Beads contain MS-compatible magnetic beads specifically designed to tolerate harsh MS sample prep conditions to improve sample quality and reproducibility for both manual and automated sample preparation methods.

Proteomic sample preparation for MS analysis is complex, resulting in challenges with reproducibility and peptide quality. For MS sample preparation methods that utilize magnetic beads, a major challenge is the compatibility of the beads with the harsh conditions required for sample processing. When exposed to extreme pH or solvents, traditional magnetic beads often clump or dissolve, resulting in increased variability.

Pierce Proteomics Magnetic Clean-up Beads have been designed for compatibility with common solvents (e.g. acetonitrile, ethanol, isopropyl alcohol, and acetone) and wide range of pH (e.g. pH 2-13) to enable fast, efficient, and reproducible removal of a wide variety of detergents, salts, and other mass spectrometry (MS)-incompatible contaminants from protein and peptide samples. The beads can be successfully used to clean up proteins derived from different sample types (e.g. purified proteins, cell lysates, tissue extracts, and biological fluids) and protein input amounts (e.g. 1 µg to 1 mg) using manual magnetic processing or automated workflows. 

EasyPep Magnetic MS Sample Prep Kits integrate optimized EasyPep sample preparation chemistry with the Proteomics Magnetic Clean-up Beads to support manual and automated magnetic bead sample processing, including the King Fisher Instrument Series. The complete kits contain pre-formulated buffers, MS-grade enzyme mix, magnetic beads for peptide clean-up, and an optimized protocol to generate MS-ready peptide samples in less than 3 hours. The kits are suitable for processing protein samples from 15–100 µg with a high yield of detergent-free peptides ready for direct LC-MS analysis without the need to SpeedVac dry and reconstitute before injection.  In addition, the peptide samples are compatible with isobaric tag (e.g., TMT/TMTpro Reagents) labeling and high pH reversed-phase fractionation.

Specifications

Product Type Magnetic MS Sample Prep Kit
Product Line EasyPep
Starting Material Protein Samples
Detection Method Mass Spectrometry
Format Kit
Quantity 20 Reactions
Form Liquid

For Research Use Only.