missing translation for 'onlineSavingsMsg'
Learn More

Contec™ DualClean™ Softwall Strip Cleaner System
GREENER_CHOICE

Designed to thoroughly and economically clean vinyl strip curtains with ease. DualClean™ Softwall Strip Cleaner System is a unique system that enables one operator to quickly clean both sides of a 4 to 12 in. plastic strip curtains in a single pass.

$651.84 - $1,085.95

Specifications

Color Code White
For Use With (Application) Enables one operator to easily and quickly clean both sides of the plastic strip curtains in a single pass
Color-coded No
View More Specs

Products 4
Catalog Number Mfr. No. Product Type Material Sterility Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Material Sterility Price Quantity & Availability  
19168808
GREENER_CHOICE
View Documents Promotion Details
Contec™
DCMD0001
Dual Clean Cover Polyester Microfiber Non-sterile
Case of 80 Each for $1,085.95
Only null left
 
19168807
GREENER_CHOICE
View Documents Promotion Details
Contec™
DCQT0001
Dual Clean Cover Polyester Knit Non-sterile
Case of 80 Each for $651.84
Only null left
 
19168805
GREENER_CHOICE
View Documents Promotion Details
Contec™
2778
Dual Clean Frame Stainless Steel (304) Non-sterile
Each for $659.43
Only null left
 
19168820
GREENER_CHOICE
View Documents Promotion Details
Contec™
DCQT0002
Dual Clean Cover Polyester Knit Sterile
Case of 80 Each for $1,085.95
Only null left
 
Description

Description

DualClean System:

The system includes stainless steel frame with two curved-surface arms, fabric covers and a QuickConnect™ handle. QuickConnect handles are interchangeable. The recommended handle for the DualClean system is cat. no. 19168806, extendable fiberglass handle. All parts are sold separately.
  • Easily and uniformly cleans both sides of the length of the strip
  • Two fabric covers available to meet cleaning needs:
    • Microfiber terrycloth removes and traps dust and other visible contaminants
    • Quiltec™ knit polyester is ideally suited to critical environments
  • Frame attaches to a wide variety of Contec handles
  • Electropolished frame is autoclavable and resistant to most chemicals and disinfectants
Specifications

Specifications

White
Enables one operator to easily and quickly clean both sides of the plastic strip curtains in a single pass
No
Videos
SDS
Documents

Documents