missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EASY-Spray™ HPLC Columns

Catalog No. 03251879
Change view
Click to view available options
Length (Metric):
150 mm
250 mm
500 mm
750 mm
Diameter (Metric):
50 μm
75 μm
150 μm
Particle Size:
2 μm
2.6 μm
3 μm
4 μm
Packing Material:
PSDVB (Polystyrene Divinylbenzene)
Spherical, Fully Porous, Ultrapure Silica
Spherical, Solid Core, Ultrapure Silica
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric) Particle Size Packing Material
03251879 150 mm 75 μm 2.6 μm Spherical, Solid Core, Ultrapure Silica
03251871 150 mm 75 μm 3 μm Spherical, Fully Porous, Ultrapure Silica
03251872 150 mm 50 μm 2 μm Spherical, Fully Porous, Ultrapure Silica
03251873 250 mm 75 μm 2 μm Spherical, Fully Porous, Ultrapure Silica
03251874 500 mm 75 μm 2 μm Spherical, Fully Porous, Ultrapure Silica
03251875 150 mm 75 μm 2 μm Spherical, Fully Porous, Ultrapure Silica
03251876 750 mm 75 μm 2 μm Spherical, Fully Porous, Ultrapure Silica
03251877 150 mm 150 μm 2 μm Spherical, Fully Porous, Ultrapure Silica
03254131 150 mm 150 μm 4 μm PSDVB (Polystyrene Divinylbenzene)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 03251879 Supplier Thermo Scientific™ Supplier No. ES911

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Robust capillary flow HPLC columns for sensitive protein, peptide and mAbs separation using the stand alone or integrated easy to use design.

Ensure robust nano and capillary flow into your LC-MS using Thermo Scientific™ EASY-Spray™ HPLC Columns. The integrated column/emitter design eliminates dead volume and is temperature-controlled for maximum reliability and performance. Rigorously tested to ensure maximum quality, these columns deliver maximum simplicity and ease-of-use. The capillary flow HPLC columns provide sensitive protein, peptide, and monoclonal antibody (mAb) separation. They give proteomics researchers more than ever before: more throughput, more sensitivity, more separation power and more ease of use.

The unique EASY-Spray design provides uncompromised performance and ease-of-use in nano and capillary LC-MS proteomics applications. Features include the following:

  • Precision positioned glass emitter
  • Integrated precision designed union
  • Integrated temperature control

Specifications

Column Format Analytical Column, EASY-Spray
Diameter (Metric) 75 μm
Length (Metric) 150 mm
Max. Pressure 11600 psi (800 bar)
Particle Size 2.6 μm
pH 2 to 9
Pore Size 150 Å
Surface Area 80 m²/g
Temperature 60°C
USP Type L26
Stationary Phase C4
Product Line Easy Spray
Packing Material Spherical, Solid Core, Ultrapure Silica
Carbon Load 2%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Technique Reversed Phase HPLC
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.