Learn More
Thermo Scientific™ EASY-Spray™ HPLC Columns
Robust capillary flow HPLC columns for sensitive protein, peptide and mAbs separation using the stand alone or integrated easy to use design.
Supplier: Thermo Scientific™ ES903
Description
Ensure robust nano and capillary flow into your LC-MS using Thermo Scientific™ EASY-Spray™ HPLC Columns. The integrated column/emitter design eliminates dead volume and is temperature-controlled for maximum reliability and performance. Rigorously tested to ensure maximum quality, these columns deliver maximum simplicity and ease-of-use. The capillary flow HPLC columns provide sensitive protein, peptide, and monoclonal antibody (mAb) separation. They give proteomics researchers more than ever before: more throughput, more sensitivity, more separation power and more ease of use.
The unique EASY-Spray design provides uncompromised performance and ease-of-use in nano and capillary LC-MS proteomics applications. Features include the following:
- Precision positioned glass emitter
- Integrated precision designed union
- Integrated temperature control
Specifications
| Analytical Column, EASY-Spray | |
| 500 mm | |
| 2 μm | |
| 100 Å | |
| 60°C | |
| C18 | |
| Spherical, Fully Porous, Ultrapure Silica | |
| Reversed Phase | |
| Reversed Phase |
| 75 μm | |
| 14500 psi (1000 bar) | |
| 2 to 8 | |
| 450 m²/g | |
| L1 | |
| Easy Spray | |
| 15% | |
| Yes |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.