missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eagle™ Floor Model Corrosives Cabinets: Two-Door Models

Provide more space to store hazardous material in your work environment. Eagle Floor Model Corrosives Cabinet can be used for storing containers up to 60 gallons in capacity.
$3,043.43 - $5,691.40
Specifications
| Certifications/Compliance | FM, NFPA, OSHA |
|---|---|
| Color | Blue |
| Material | Steel |
| Product Type | Corrosives Cabinet |
| For Use With (Application) | Flammables |
Alert:
Not recommended for use with sulfuric, nitric, or hydrochloric acid.
| Catalog Number | Mfr. No. | Door Style | Load Bearing Capacity | Dimensions (D x W x H) Interior | No. of Shelves | Price | Quantity & Availability | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Door Style | Load Bearing Capacity | Dimensions (D x W x H) Interior | No. of Shelves | Price | Quantity & Availability | ||
171562E
![]() |
Eagle™
CRA62 |
Manual-close | 60 gal. | 28.25 x 28.25 x 62 in. | 2 |
Each for $5,144.31
|
|
||
171562B
![]() |
Eagle™
CRA32 |
Manual-close | 30 gal. | 15 x 40 x 41 in. | 1 |
Each for $3,043.43
|
|
||
19120717
![]() |
Eagle™
CRA3010X |
Self-close | 30 gal. | 15 x 40 x 41 in. | 1 |
Each for $3,495.86
|
|
||
19120718
![]() |
Eagle™
CRA4510 |
Self-close | 45 gal. | 15 x 40 x 62 in. | 2 |
Each for $4,933.67
|
|
||
19120719
![]() |
Eagle™
CRA6010X |
Self-close | 60 gal. | 28.25 x 28.25 x 62 in. | 2 |
Each for $5,691.40
|
|
||
171562D
![]() |
Eagle™
CRA47X |
Manual-close | 45 gal. | 15 x 40 x 62 in. | 2 |
Each for $4,548.46
|
|
||
Description
Provide more space to store hazardous material in your work environment. Eagle Floor Model Corrosives Cabinet can be used for storing containers up to 60 gallons in capacity.
- Suitable for storage of flammable and nonflammable acids and corrosive liquids
- Rugged 18-gauge steel cabinets coated with tough blue epoxy powder on interior and exterior surfaces, including shelves, for extra protection against corrosive acids and caustics
- Coated surfaces are superior to epoxy- or enamel-painted surfaces in case of spill or splash
- Doors clearly labeled CAUTION—CORROSIVES
- High-density polyethylene trays for shelves and bottom with drip edge contains liquid spills; removable for easy cleaning
Certifications
FM approved.
Compliance
Meet NFPA Code 30 and OSHA requirements.
Specifications
| FM, NFPA, OSHA | |
| Steel | |
| Flammables | |
| 2 |
| Blue | |
| Corrosives Cabinet | |
| HDPE shelves and bottom trays with drip edge | |
| Epoxy |