missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eagle™ Floor Model Corrosives Cabinets: Two-Door Models

Description
Provide more space to store hazardous material in your work environment. Eagle Floor Model Corrosives Cabinet can be used for storing containers up to 60 gallons in capacity.
- Suitable for storage of flammable and nonflammable acids and corrosive liquids
- Rugged 18-gauge steel cabinets coated with tough blue epoxy powder on interior and exterior surfaces, including shelves, for extra protection against corrosive acids and caustics
- Coated surfaces are superior to epoxy- or enamel-painted surfaces in case of spill or splash
- Doors clearly labeled CAUTION—CORROSIVES
- High-density polyethylene trays for shelves and bottom with drip edge contains liquid spills; removable for easy cleaning
Certifications
FM approved.
Compliance
Meet NFPA Code 30 and OSHA requirements.
Specifications
Specifications
| Dimensions (D x W x H) Interior | 28.25 x 28.25 x 62 in. |
| Product Type | Corrosives Cabinet |
| Width (Metric) Interior | 71.75 cm |
| Weight (Metric) | 164 kg |
| Width (Metric) Exterior | 79.3 cm |
| No. of Shelves | 2 |
| Depth (Metric) Exterior | 79.3 cm |
| Height (Metric) Exterior | 165.1 cm |
| No. of Doors | 2 |
| Height (Metric) Interior | 157.48 cm |
| Show More |