missing translation for 'onlineSavingsMsg'
Learn More

Eagle™ Floor Model Corrosives Cabinets: Two-Door Models
GREENER_CHOICE

Catalog No. 19120719
Change view
Click to view available options
Door Style:
Manual-close
Self-close
Load Bearing Capacity:
30 gal.
45 gal.
60 gal.
Dimensions (D x W x H) Interior:
28.25 x 28.25 x 62 in.
15 x 40 x 41 in.
15 x 40 x 62 in.
No. of Shelves:
1
2
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Door Style Load Bearing Capacity Dimensions (D x W x H) Interior No. of Shelves
19120719 Self-close 60 gal. 28.25 x 28.25 x 62 in. 2
171562E Manual-close 60 gal. 28.25 x 28.25 x 62 in. 2
171562B Manual-close 30 gal. 15 x 40 x 41 in. 1
19120717 Self-close 30 gal. 15 x 40 x 41 in. 1
19120718 Self-close 45 gal. 15 x 40 x 62 in. 2
171562D Manual-close 45 gal. 15 x 40 x 62 in. 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options

Not recommended for use with sulfuric, nitric, or hydrochloric acid.

Catalog No. 19120719 Supplier Eagle™ Supplier No. CRA6010X

Not recommended for use with sulfuric, nitric, or hydrochloric acid.

Provide more space to store hazardous material in your work environment. Eagle Floor Model Corrosives Cabinet can be used for storing containers up to 60 gallons in capacity.

Provide more space to store hazardous material in your work environment. Eagle Floor Model Corrosives Cabinet can be used for storing containers up to 60 gallons in capacity.

  • Suitable for storage of flammable and nonflammable acids and corrosive liquids
  • Rugged 18-gauge steel cabinets coated with tough blue epoxy powder on interior and exterior surfaces, including shelves, for extra protection against corrosive acids and caustics
  • Coated surfaces are superior to epoxy- or enamel-painted surfaces in case of spill or splash
  • Doors clearly labeled CAUTION—CORROSIVES
  • High-density polyethylene trays for shelves and bottom with drip edge contains liquid spills; removable for easy cleaning

Certifications

FM approved.

Compliance

Meet NFPA Code 30 and OSHA requirements.

Specifications

Product Type Corrosives Cabinet
Dimensions (D x W x H) Interior 28.25 x 28.25 x 62 in.
Load Bearing Capacity 60 gal.
Door Style Self-close
Width (Metric) Exterior 79.3 cm
Depth (Metric) Exterior 79.3 cm
Weight (Metric) 165 kg
Height (Metric) Exterior 165.1 cm
Width (Metric) Interior 71.75 cm
No. of Shelves 2
Height (Metric) Interior 157.48 cm
Depth (Metric) Interior 71.7 cm
No. of Doors 2
Certifications/Compliance FM, NFPA, OSHA
For Use With (Application) Flammables
Weight (English) 364 lb.
Depth (English) Exterior 31.25 in.
Width (English) Exterior 31.25 in.
Height (English) Interior 62 in.
Width (English) Interior 28.25 in.
Includes HDPE shelves and bottom trays with drip edge
Material Steel
Height (English) Exterior 65 in.
Depth (English) Interior 28.25 in.
Surface Coating Epoxy
Color Blue
Show More Show Less