missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-PAGE™ Midi Protein Gels, 3.7 mm

Catalog No. p-7237830
Encompass
Change view
Click to view available options
Wells:
48-well
96-well
Gel Percentage:
6%
8%
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Wells Gel Percentage
EP09606 96-well 6%
EP04808 48-well 8%
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EP09606 Supplier Invitrogen™ Supplier No. EP09606
Only null left

Includes

8 E-PAGE™ 96 gels, 4.5mL E-PAGE™ Loading Buffer 1 (4X), 1 Butterfly Opener, 1 E-PAGE™ Blotting Pad

E-PAGE™ Gels are self-contained, pre-cast gels that include a gel matrix and electrodes packaged inside a disposable cassette.

E-PAGE Gels are self-contained, pre-cast gels that include a gel matrix and electrodes packaged inside a disposable cassette. Protein separation on an E-PAGE Gel requires an EBase Integrated Device, a compact and automated platform designed for medium- and high-throughput electrophoresis.

Each E-PAGE 8% 48-well gel contains 48 sample lanes and 4 marker lanes with a 3.2-cm separation length. E-PAGE 48-well gels can be loaded with a 12- or 8-channel pipettor or 4- or 8-tip robotic system (Figure 1).

Each E-PAGE 6% 96-well gel contains 96 sample lanes and 8 marker lanes with a 1.6-cm separation length. The well openings of the cassette are compatible with the standard 96-well plate format and can be conveniently loaded with a multichannel pipettor or 8-, 12-, or 96-tip liquid-handling robotic device (Figure 2).

Both E-PAGE 48 and 96 cassettes can be opened to remove the gel for downstream staining or blotting (Figures 3 and 4). In addition, each cassette is labeled with an individual barcode to facilitate identification of the gel using commercial barcode readers.

More about the E-PAGE precast gel system
The Invitrogen E-PAGE precast gel system is designed for fast, mess-free medium- and high-throughput protein electrophoresis. It consists of the E-PAGE precast gels, E-Base Integrated devices for running the gels, and free E-Editor 2.0 software for image analysis.

The all-in-one E-Gel Power Snap Plus Electrophoresis System is designed for fast and convenient high-throughput gel electrophoresis. With dry precast E-PAGE gel technology, you can run protein samples in as little as 14 minutes. The device arrives with pre-programmed protocols for both E-PAGE gel types.

Order Info

Shipping Condition: Room Temperature

Specifications

Gel Percentage 6%
Gel Size Midi
Gel Thickness 3.7 mm
Gel Type E-PAGE
Sample Loading Volume Up to 20 μL
Separation Range 10 to 300 kDa
Separation Type Denaturing
Wells 96-well
Mode of Separation Molecular Weight
Quantity 8 gels (1 kit)
Shipping Condition Room Temperature
Storage Requirements Store at room temperature, do not allow the temperature to drop below 4°C or rise above 40°C
Length (Metric) 8 cm
Width (Metric) 13 cm
Product Line E-PAGE
For Use With (Equipment) E-BASE
Show More Show Less