missing translation for 'onlineSavingsMsg'
Learn More

(E)-alpha-Methylstilbene, 98%

Catalog No. p-7022528
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1535106 5 g
AAA1535114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1535106 Supplier Thermo Scientific Chemicals Supplier No. A1535106
Only null left

CAS: 833-81-8 | C15H14 | 194.277 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 833-81-8
Molecular Formula C15H14
Molecular Weight (g/mol) 194.277
MDL Number MFCD00026343
InChI Key OVZXISBUYCEVEV-OUKQBFOZSA-N
Synonym 1,2-diphenylpropene, e-alpha-methylstilbene, .alpha.-methylstilbene, 1,2-diphenyl-1-propene, 1-methyl-1,2-diphenylethene, stilbene, .alpha.-methyl, 1-propene, 1,2-diphenyl, 1-methyl-2-phenylvinyl benzene, trans-alpha-methylstilbene, chembl14773
PubChem CID 1549166
IUPAC Name [(E)-1-phenylprop-1-en-2-yl]benzene
SMILES CC(=CC1=CC=CC=C1)C2=CC=CC=C2

Specifications

Melting Point 79°C to 82°C
Density 0.986
Quantity 5 g
Beilstein 1906425
Formula Weight 194.28
Percent Purity 98%
Chemical Name or Material (E)-alpha-Methylstilbene
EINECSNumber 212-300-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only