missing translation for 'onlineSavingsMsg'
Learn More

(E)-1,2-Bis(2-naphthyl)-1,2-bis(1,1,3,3,5,5,7,7-octaethyl-1,2,3,5,6,7-hexahydro-s-indacen-4-yl)disilene 97.0+%, TCI America™

Catalog No. B442250MG
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B442250MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B442250MG Supplier TCI America Supplier No. B442250MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1253900-41-2
Molecular Formula C76H104Si2
Molecular Weight (g/mol) 1073.838
InChI Key JCRYFUKOTHHJSV-JKNXPUSASA-N
Synonym (Eind)(2-Naph)Si=Si(2-Naph)(Eind)
PubChem CID 91972172
IUPAC Name (E)-naphthalen-2-yl-[naphthalen-2-yl-(1,1,3,3,5,5,7,7-octaethyl-2,6-dihydro-s-indacen-4-yl)silylidene]-(1,1,3,3,5,5,7,7-octaethyl-2,6-dihydro-s-indacen-4-yl)silane
SMILES CCC1(CC(C2=C(C3=C(C=C21)C(CC3(CC)CC)(CC)CC)[Si](=[Si](C4=CC5=CC=CC=C5C=C4)C6=C7C(=CC8=C6C(CC8(CC)CC)(CC)CC)C(CC7(CC)CC)(CC)CC)C9=CC1=CC=CC=C1C=C9)(CC)CC)CC

Specifications

Melting Point 285°C
Quantity 50 mg
Formula Weight 1073.84
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (E)-1,2-Bis(2-naphthyl)-1,2-bis(1,1,3,3,5,5,7,7-octaethyl-1,2,3,5,6,7-hexahydro-s-indacen-4-yl)disil