accessibility menu, dialog, popup

UserName

CAS RN 1253900-41-2

IUPAC Name: 1,2-bis(naphthalen-2-yl)-1,2-bis(1,1,3,3,5,5,7,7-octaethyl-1,2,3,5,6,7-hexahydro-s-indacen-4-yl)disilene
Synonyms: 1,2-bis(naphthalen-2-yl)-1,2-bis(1,1,3,3,5,5,7,7-octaethyl-2,6-dihydro-s-indacen-4-yl)disilene
Molecular Weight (g/mol): 1073.84
Molecular Formula: C76H104Si2
InChi Key: JCRYFUKOTHHJSV-UHFFFAOYSA-N
SMILES: CCC1(CC)CC(CC)(CC)C2=C(C3=C(C=C12)C(CC)(CC)CC3(CC)CC)[Si](C1=CC=C2C=CC=CC2=C1)=[Si](C1=CC=C2C=CC=CC2=C1)C1=C2C(=CC3=C1C(CC)(CC)CC3(CC)CC)C(CC)(CC)CC2(CC)CC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results