missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ DZNep HCl
Description
DZNep-RMU is an Ancillary Material Grade version of product 3-Deazaneplanocin A hydrochloride (Cat. No. 4703). Suitable for use as an ancillary reagent in the further manufacturing of cell therapies. Find out more about our Ancillary Material Grade product range.
Specifications
Specifications
| Chemical Name or Material | (1 S,2 R,5 R)-5-(4-Amino-1 H-imidazo[4,5-c]pyridin-1-yl)-3-(hydroxymethyl)-3-cyclopentene-1,2-diol hydrochloride |
| CAS | 120964-45-6 |
| Quantity | 5 mg |
| Target | EZH2 Inhibitor |
| Molecular Formula | C12H14N4O3.HCl |
| Purity | 0.98 |