missing translation for 'onlineSavingsMsg'
Learn More

Dysprosium AA Standard, 1000 ppm Dy in 3% HNO3, Ricca Chemical

Catalog No. ADY1KN100
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ADY1KN100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ADY1KN100 Supplier Ricca Chemical Company Supplier No. ADY1KN100

Chemical Identifiers

CAS 1308-87-8 , 7697-37-2 , 1308-87-8
Molecular Formula Dy2O3
Molecular Weight (g/mol) 373.00
MDL Number MFCD00010986
InChI Key GEZAXHSNIQTPMM-UHFFFAOYSA-N
IUPAC Name didysprosium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Dy+3].[Dy+3]

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm Dy
CAS Min % 93.0, 4.62, 0.21
CAS Max % 97.0, 4.81, 0.22
Equivalence 1 mL = 1 mg Dy
Physical Form Liquid
Color Colorless
pH < 2
Melting Point Approximately 0°C
Boiling Point Approximately 100°C
For Use With (Equipment) Atomic Absorption Spectrophotometer
Solubility Miscible with water
Solution Type Calibration Standard
Type Single-element Standard Solution
Quantity 100 mL
Grade Laboratory
Chemical Name or Material Dysprosium AA Standard
Show More Show Less