missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Dynole 34-2

Catalog No. 422210
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
422210 10 mg
422250 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 422210 Supplier Tocris Bioscience™ Supplier No. 4222/10

Dynamin I inhibitor

Dynamin I inhibitor (IC50 = 1.3 μM). Inhibits receptor-mediated endocytosis (RME) (IC50 = 5.0 μM).

Chemical Identifiers

CAS 1128165-88-7
Molecular Formula C25H36N4O

Specifications

Quantity 10 mg
Formula Weight 408.58
Purity >99%
Molecular Weight (g/mol) 408.59
Chemical Name or Material Dynamin Inhibitors
Recommended Storage Store at +4°C