missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DyNAmo HS SYBR Green qPCR Kit

Catalog No. F410L
Change view
Click to view available options
No. of Reactions:
2500 Reactions
500 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
F410L 500 Reactions
F410XL 2500 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F410L Supplier Thermo Scientific™ Supplier No. F410L
Only null left

Contains a modified Thermus brockianus DNA polymerase and all the other reagents needed for qPCR.

Only template DNA and PCR primers need to be added by the user. The modified DNA polymerase in this kit incorporates a non-specific DNA-binding domain that lends physical stability to the polymerase-DNA complex. ROX passive reference dye is also supplied with the kit in a separate tube for instruments that require ROX normalization.

  • Strong specificity: DyNAmo HS SYBR Green master mix minimizes the formation of unwanted primer-dimers and extension of non-specifically bound primers during reaction setup
  • Highly sensitive detection: Optimized master mix allows reproducible detection of low-copy number templates
  • Broad dynamic range: DyNAmo HS SYBR Green qPCR Kit detects as few as one copy of the target gene and achieves a dynamic range of up to eight orders of magnitude
  • Convenient to use: 2X master mix, one tube only; ROX passive reference dye provided in a separate vial

Specifications

Concentration 2X (qPCR Mix), 50X (Dye)
Content And Storage Contains:
• 5 x1 mL DyNAmo HS SYBR Green qPCR Mix (2X)
• 250 μL 50X ROX dye, 25μM

Store at -20°C
Detection Method SYBR
Format Tube
GC-Rich PCR Performance High
PCR Method qPCR
Polymerase Tbr DNA polymerase
Reaction Speed Standard
For Use With (Equipment) 7000 System, 7300 System, 7500 Fast System, 7500 System, 7700 System, 7900HT Fast System, 7900HT System, BioRad CFX96™, QuantStudio™, ViiA™ 7 System, Bio-Rad CFX384 Touch™, Roche LightCycler 480, 5700 Sequence Detection System, StepOnePlus™ System, Eppendorf MasterCycler RealPlex, Corbett RotorGene, StepOne™ System, Stratagene Mx4000, Stratagene Mx3005P, Stratagene Mx3000P, MJ Opticon, MJ Chromo4, Cepheid SmartCycler
Kit Type DyNAmo HS SYBR Green qPCR Kit
Product Line DyNAmo
Product Type Real Time PCR SYBR Master Mix
Quantity 500 reactions
Sufficient For 500 Reactions of 20 μL Each, 200 Reactions of 50 μL Each
For Use With (Application) Gene Expression
No. of Reactions 500 Reactions
Sample Type DNA, dsDNA
Volume 5 x 1 mL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.