missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DynaLight™ Substrate with RapidGlow™ Enhancer

Catalog No. 4475406
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4475406 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4475406 Supplier Invitrogen™ Supplier No. 4475406
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A ready-to-use chemiluminescent substrate formulation optimized to achieve faster results in solution-based immunoassays. Thermo Scientific DynaLight Substrate with RapidGlow Enhancer provides a robust chemiluminescent reaction when catalyzed by alkaline

When combined with DynaLight Trigger Solution, DynaLight Substrate with RapidGlow Enhancer delivers maximum signal within seconds. The environmentally-friendly DynaLight Trigger Solution provides maximum signal within 1 second of addition, with a longer half-life to its ground state than other chemiluminescent Flash technologies, allowing ample time to collect data reliably.

  • Faster kinetics and higher signal levels than colorimetric, fluorescent, or other chemiluminescent chemistries
  • Sensitive attomole detection of purified alkaline phosphatase enzyme
  • Optimized formulation for both microplate and bead-based assays
  • Substrate stability of over 1 year at 4°C
  • Compatibility with automated platforms with trigger solution/positions already in place
  • Includes 1,2-dioxetane chemiluminescent substrate and a polymeric enhancer that enables ultra-sensitive immunoassay detection by alkaline phosphatase label

Specifications

Product Type ELISA Substrate
Quantity 100 mL
Detection Method Chemiluminescence
Assay Sensitivity 1 pg/mL
Form Liquid
Format Bottle
For Use With (Equipment) Luminometer (Microplate)
High-throughput Compatibility High-throughput Compatible
Product Line DynaLight, NovaBright, RapidGlow
Sample Type Cell Cultures, Whole Blood, Serum
Substrate Properties Chemical Substrate
Substrate Type Alkaline Phosphatase
Test Time 2 min.
Content And Storage 1 amber bottle — Store at 2 to 8°C
Shipping Condition Wet Ice
Show More Show Less

For Research Use Only. Not for human or animal therapeutic or diagnostic use.