Learn More
DWK Life Sciences DURAN™ GLS 80™ Laboratory Wide Neck Glass Bottles w/Screw-Cap and Pour Ring
The 80 mm wide outer diameter of the bottle neck permits easy filling and pouring out of powders and viscous substances.
Supplier:  DWK Life Sciences 1113951
 View more versions of this product
Catalog No. 09841227
Only null left
Description
Description
  • Typical applications: storage, transport, safekeeping and sampling of substances, easy to use with granulated material, powders and viscous media, sampling of hot media
Specifications
Chemical Identifiers
Clear
Screw-cap and pouring ring
Glass
Blue
Specifications
20 L
Clear
Glass
484 mm
288 mm
20 L
Screw-cap and pouring ring
Blue
Laboratory Bottle
Product Suggestions
Product Suggestions