missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ GLS 80™ Laboratory Wide Neck Glass Bottles w/Screw-Cap and Pour Ring
The 80 mm wide outer diameter of the bottle neck permits easy filling and pouring out of powders and viscous substances.
Supplier: DWK Life Sciences 1113950
Description
- Typical applications: storage, transport, safekeeping and sampling of substances, easy to use with granulated material, powders and viscous media, sampling of hot media
Chemical Identifiers
| Clear | |
| Screw-cap and pouring ring | |
| Glass | |
| Blue |
Specifications
| 10 L | |
| Clear | |
| Glass | |
| 389 mm | |
| 227 mm |
| 10 L | |
| Screw-cap and pouring ring | |
| Blue | |
| Laboratory Bottle |