Learn More
DuPont™ Tychem™ 2000 Coverall (all)
Flexible and durable light liquid splash protection
$169.40 - $235.46
Specifications
Gender Unisex
Color Yellow
Ankle Style Open
Closure Type Zipper
View More Specs
Products 10
Catalog Number Mfr. No. Seam Type Size Price Quantity  
Catalog Number Mfr. No. Seam Type Size Price Quantity  
19-812-695
View Documents
DuPont™
QC120SYLMD001200
Serged Medium
Case of 12 Each for $180.48
Only null left
18999593E
View Documents
DuPont™
QC120SYL4X001200
Serged 4X-Large
Case of 12 Each for $205.59
Only null left
19-812-696
View Documents
DuPont™
QC120SYLXL001200
Serged X-Large
Case of 12 Each for $180.48
Only null left
18999593D
View Documents
DuPont™
QC120SYL3X001200
Serged 3X-Large
Case of 12 Each for $190.32
Only null left
18999593A
View Documents
DuPont™
QC120SYLLG001200
Serged Large
Case of 12 Each for $169.40
Only null left
18-999-593C
View Documents
DuPont™
QC120SYL2X001200
Serged 2X-Large
Case of 12 Each for $199.80
Only null left
19-041-747
View Documents
DuPont™
QC120BYLLG001200
Bound Large
Case of 12 Each for $209.52
Only null left
19-041-760
View Documents
DuPont™
QC120BYL2X001200
Bound 2X-Large
Case of 12 Each for $222.12
Only null left
19-715-319
View Documents
DuPont™
QC120BYL3X001200
Bound 3X-Large
Case of 12 Each for $235.46
Only null left
18-999-593F
View Documents
DuPont™
QC120SYL5X001200
Serged 5X-Large
Case of 12 Each for $222.01
Only null left
Description
Description
Tychem™ QC is polyethylene-coated Tyvek™ fabric fabric provides at least 30 minutes of protection against 42 chemical challenges
  • Laydown collar
  • Open wrists
  • Open ankles
  • Yellow for high visibility
  • 10 mil thickness
QC 120B
  • Tightly sewn seam is covered with garment fabric to reinforce seam and reduce potential for particle penetration
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
QC 120S
  • Multiple interlocking threads are sewn around the raw edges of garment material to create a strong, stress-resistant seams
QC 120T
  • Taped seams provide strong chemical resistance against heavy liquid splashes
  • A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
Specifications
Specifications
Unisex
Yellow
Open
Zipper
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'