missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tychem™ 5000 Hooded Coverall
Great tear resistance
$584.45 - $637.35
Specifications
| Gender | Unisex |
|---|---|
| Material | Tychem |
| Color | Tan |
| Ankle Style | Elastic |
| Cuff Style | Elastic |
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|
29-633-813
|
DuPont™
C3127TTN2X000600 |
2X-Large |
Case of 6 Each for $621.66
|
|
|||||
|
19-087104A
|
DuPont™
C3127TTNLG000600 |
Large |
Case of 6 Each for $630.92
|
|
|||||
|
29-663-702
|
DuPont™
C3127TTNMD000600 |
Medium |
Case of 6 Each for $637.35
|
|
|||||
|
29-663-703
|
DuPont™
C3127TTNXL000600 |
X-Large |
Case of 6 Each for $584.45
|
|
|||||
Description
- Zipper features a semi-automatic locking slider allowing the puller to stay down and in a locked position
- Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
- Attached hood with elastic around face opening.
- Strong zipper for tough, flexible closure
- Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
- Elastic opening for tighter fit at wrist
- Elastic opening for tighter fit at ankle
- Manufactured under specifications that do not contain natural rubber latex
- Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.
Specifications
| Unisex | |
| Tan | |
| Elastic | |
| Standard | |
| Taped | |
| Chemical handling, petrochemical, hazardous materials, waste cleanup, fire departments, industrial hazmat teams, utilities and domestic preparedness |
| Tychem | |
| Elastic | |
| Hooded | |
| Zipper | |
| Standard fit hood, elastic wrist and ankle; Multilayer composite barrier film laminated to a durable Polypropylene substrate |