missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tychem™ 5000 Hooded Coverall

Catalog No. 29633813
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
29633813 2X-Large
19087104A Large
29663702 Medium
29663703 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 29633813 Supplier DuPont™ Supplier No. C3127TTN2X000600

Great tear resistance

  • Zipper features a semi-automatic locking slider allowing the puller to stay down and in a locked position
  • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
  • Attached hood with elastic around face opening.
  • Strong zipper for tough, flexible closure
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle
  • Manufactured under specifications that do not contain natural rubber latex
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.

Specifications

Size 2X-Large
Gender Unisex
Material Tychem
Color Tan
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Standard
Closure Type Zipper
Seam Type Taped
Description Standard fit hood, elastic wrist and ankle; Multilayer composite barrier film laminated to a durable Polypropylene substrate
Size (English) Inseam 30 in.
Size (Metric) Chest 114.93 to 123.82 cm
For Use With (Application) Chemical handling, petrochemical, hazardous materials, waste cleanup, fire departments, industrial hazmat teams, utilities and domestic preparedness
Length (English) Sleeve 37 in.
Length (Metric) Sleeve 93.98 cm
Width (English) Chest 29.25 in.
Height (English) 6 to 6.4 ft.
Height (Metric) 1.82 to 1.95 m
Show More Show Less