missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Dulcitol, 99+%
CAS: 608-66-2 | C6H14O6 | 182.17 g/mol
$108.08 - $817.72
Chemical Identifiers
| CAS | 608-66-2 |
|---|---|
| Molecular Formula | C6H14O6 |
| Molecular Weight (g/mol) | 182.17 |
| MDL Number | MFCD00064288 |
| InChI Key | FBPFZTCFMRRESA-GUCUJZIJSA-N |
| Synonym | dulcitol, galactitol, dulcite, d-galactitol, dulcose, euonymit, melampyrin, melampyrit, d-dulcitol, 2r,3s,4r,5s-hexane-1,2,3,4,5,6-hexaol |
| PubChem CID | 11850 |
| ChEBI | CHEBI:16813 |
| IUPAC Name | (2R,3S,4R,5S)-hexane-1,2,3,4,5,6-hexol |
| SMILES | C(C(C(C(C(CO)O)O)O)O)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC117700250
|
Thermo Scientific Chemicals
117700250 |
25 g | Glass bottle |
Each for $108.08
|
|
||||
|
AC117701000
|
Thermo Scientific Chemicals
117701000 |
100 g | Glass bottle |
Each for $267.67
|
|
||||
|
AC117705000
|
Thermo Scientific Chemicals
117705000 |
500 g | Glass bottle |
Each for $817.72
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C6H14O6 | |
| MFCD00064288 | |
| dulcitol, galactitol, dulcite, d-galactitol, dulcose, euonymit, melampyrin, melampyrit, d-dulcitol, 2r,3s,4r,5s-hexane-1,2,3,4,5,6-hexaol | |
| CHEBI:16813 | |
| C(C(C(C(C(CO)O)O)O)O)O |
Specifications
| 608-66-2 | |
| White | |
| Authentic | |
| C6H14O6 | |
| 01, 544 | |
| dulcitol, galactitol, dulcite, d-galactitol, dulcose, euonymit, melampyrin, melampyrit, d-dulcitol, 2r,3s,4r,5s-hexane-1,2,3,4,5,6-hexaol | |
| FBPFZTCFMRRESA-GUCUJZIJSA-N | |
| (2R,3S,4R,5S)-hexane-1,2,3,4,5,6-hexol | |
| 11850 | |
| 182.17 | |
| Powder |
| 188°C to 189°C | |
| 275°C to 280°C (1.0 mmHg) | |
| 99% min. (ELSD) (HPLC) | |
| MFCD00064288 | |
| 15, 4362 | |
| (3% in water) Colorless | |
| C(C(C(C(C(CO)O)O)O)O)O | |
| 182.17 | |
| CHEBI:16813 | |
| 99+% | |
| Dulcitol, 99+% |
Safety and Handling
EINECSNumber : 210-165-2
TSCA : TSCA
RUO – Research Use Only