missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Dulcitol, 97%

CAS: 608-66-2 | C6H14O6 | 182.172 g/mol

Supplier:  Thermo Scientific Chemicals A1840236

 View more versions of this product

Catalog No. AAA1840236


Only null left

Description

Description

Dulcitol is important in studies related to cataract formation, hepatosplenμly and mental retardation. It is used as a substrate to identify, differentiate and characterize galactitol dehydrogenase(s). It is also used as a reference compound in analytical procedures developed to analyze sugar alcohols in plants and food.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dulcitol is important in studies related to cataract formation, hepatosplenµly and mental retardation. It is used as a substrate to identify, differentiate and characterize galactitol dehydrogenase(s). It is also used as a reference compound in analytical procedures developed to analyze sugar alcohols in plants and food.

Solubility
Soluble in water (30g/L at 14°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. Store under dry inert gas. It is hygroscopic in nature.
Specifications

Specifications

Dulcitol
187°C to 191°C
275°C to 280°C (1 mmHg)
MFCD00064288
1721903
14,4332
Soluble in water (30g/L at 14°C).
C(C(C(C(C(CO)O)O)O)O)O
182.172
CHEBI:16813
97%
608-66-2
1.47
C6H14O6
500 g
Hygroscopic
dulcitol, galactitol, dulcite, d-galactitol, dulcose, euonymit, melampyrin, melampyrit, d-dulcitol, 2r,3s,4r,5s-hexane-1,2,3,4,5,6-hexaol
FBPFZTCFMRRESA-GUCUJZIJSA-N
(2R,3S,4R,5S)-hexane-1,2,3,4,5,6-hexol
11850
182.17

Chemical Identifiers

608-66-2
182.172
FBPFZTCFMRRESA-GUCUJZIJSA-N
11850
(2R,3S,4R,5S)-hexane-1,2,3,4,5,6-hexol
C6H14O6
MFCD00064288
dulcitol, galactitol, dulcite, d-galactitol, dulcose, euonymit, melampyrin, melampyrit, d-dulcitol, 2r,3s,4r,5s-hexane-1,2,3,4,5,6-hexaol
CHEBI:16813
C(C(C(C(C(CO)O)O)O)O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 210-165-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only