missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ dTRIM 24
Description
Degrader (PROTAC®) composed of the TRIM24 ligand IACS-9571 conjugated to a VHL ligand. Displays dose- and time-dependent degradation of TRIM24 in cells, with maximum degradation achieved at 5 μM. Degradation phenotypically mirrors genetic deletion of TRIM24. Demonstrates antiproliferative effects in MOLM-13 cells. Cell permeable.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| CAS | 2170695-14-2 |
| Quantity | 5 mg |
| Molecular Formula | C55H68N8O13S2 |
| Formula Weight | 1113.3 |
| Product Type | Degrader |
| Purity | 98% |