missing translation for 'onlineSavingsMsg'
Learn More

dsRNA Mouse anti-Virus, Clone: 3G1, MilliporeSigma™

Mouse Monoclonal Antibody

$284.60 - $731.13

Specifications

Antigen dsRNA
Clone 3G1
Concentration 0.5 mg/mL
Applications ELISA, Immunomicroscopy, Immunofluorescence, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MABF199625
View Documents
MilliporeSigma
MABF199625UL
25 μL
Each for $284.60
Only null left
 
MABF1996100
View Documents
MilliporeSigma
MABF1996100UL
100 μL
Each for $731.13
Only null left
 
Description

Description

dsRNA Monoclonal antibody specifically detects dsRNA in Virus samples. It is validated for ELISA, Immunomicroscopy, Immunofluorescence, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation.
Specifications

Specifications

dsRNA
0.5 mg/mL
Monoclonal
Purified
RUO
Purified mouse monoclonal antibody IgM in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
IgM κ
Ligatrap™
3G1
ELISA, Immunomicroscopy, Immunofluorescence, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation
Unconjugated
Mouse
Virus
Concentrated supernatant from C6/36 cell cultures infected with Palm Creek virus
Primary
Stable for 1 year at +2°C to +8°C from date of receipt.
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'