missing translation for 'onlineSavingsMsg'
Learn More
Learn More
dsRNA Mouse anti-Virus, Clone: 3G1, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABF1996100UL
Description
dsRNA Monoclonal antibody specifically detects dsRNA in Virus samples. It is validated for ELISA, Immunomicroscopy, Immunofluorescence, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation.Specifications
| dsRNA | |
| Monoclonal | |
| 0.5 mg/mL | |
| Purified mouse monoclonal antibody IgM in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
| Concentrated supernatant from C6/36 cell cultures infected with Palm Creek virus | |
| 100 μL | |
| Primary | |
| Stable for 1 year at +2°C to +8°C from date of receipt. | |
| IgM κ |
| ELISA, Immunomicroscopy, Immunofluorescence, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation | |
| 3G1 | |
| Unconjugated | |
| Mouse | |
| Ligatrap™ | |
| RUO | |
| Virus | |
| Purified |