missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dry-Ready™ RT-qPCR Kit

Catalog No. A40006906
Change view
Click to view available options
Quantity:
200 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A40006906 200 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A40006906 Supplier Invitrogen™ Supplier No. A40006906

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Experience the simplicity and efficiency of the air drying process by gently heating materials at moderate temperatures in a simple ventilated oven.

The Invitrogen Dry-Ready RT-qPCR Kit is an innovative solution designed for preparation of hot air-dried RT-qPCR assays. A simple and fast air-drying method surpasses traditional lyophilization in both speed and simplicity, while also enabling assay storage and shipment at room temperature.

The kit includes Dry-Ready Platinum II DNA Polymerase, Dry-Ready SuperScript III Flash Reverse Transcriptase, Dry-Ready RNaseOUT Recombinant Ribonuclease Inhibitor, Dry-Ready dNTP Mix, 10X Dry-Ready Reaction Buffer with exipients, and MgCl2 for a suite of essential components for one-step RT-qPCR applications.

The Dry-Ready RT-qPCR Kit represents a significant advancement in one-step RT-qPCR technology, offering a solution that combines innovation with reliability. Features include:

  • Complete set of components for RT-qPCR assay drying and downstream applications
  • Dry-Ready SuperScript III Flash Reverse Transcriptase enables reverse transcription in just one minute
  • Dry-Ready Platinum II DNA Polymerase helps ensure high reliability and increased tolerance to inhibitors
  • Enhanced stability, specifically optimized for air drying applications

Air-drying technology

Utilizing this kit is straightforward and efficient: users need only mix the RT-qPCR assay components and aliquot into single reaction volumes. The mixture is then dried in an oven. This uncomplicated process results in a stable assay that can be stored at room temperature for at least 12 months without loss of efficiency.

By incorporating all these features, the Dry-Ready RT-qPCR Kit stands as an innovative solution in the field of molecular diagnostics and research. Its unique combination of enzyme stability, ease of use, and long-term storage capability at room temperature positions it as a valuable asset for laboratories looking to advance their RT-qPCR capabilities while streamlining their processes.

Specifications

Content And Storage

•Dry-Ready™ Platinum™ II DNA Polymerase, 24 μL
•Dry-Ready™ SuperScript™ III Flash Reverse transcriptase, 20 μL
•Dry-Ready™ RNaseOUT™, 40 μL
•Dry-Ready™ dNTP Mix, 80 μL
•MgCl2, 40 μL
•Store at -20°C.

Detection Method qPCR, RT-PCR
Form Liquid
GC-Rich PCR Performance High
PCR Method 1-step RT-PCR
Polymerase Platinum™ II Taq Hot-Start DNA Polymerase
Reaction Speed Fast
Label or Dye Unlabeled
No. of Tests 200 Reactions
Product Type RT-qPCR Kit
Quantity 200 Reactions
Reverse Transcriptase SuperScript™ III Flash Reverse Transcriptase
Ribonuclease H Activity Reduced
Shipping Condition Dry Ice
For Use With (Application) Real Time PCR (qPCR), RT-PCR
Hot Start Built-In Hot Start
No. of Reactions 200 Reactions
Sample Type RNA & DNA
Show More Show Less

For Research Use Only.