missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DRIERITE™, Non-Indicating, 8 Mesh, J.T. Baker™
$7151.00 - $7430.22
Chemical Identifiers
| CAS | 7778-18-9 |
|---|---|
| Molecular Formula | CaO4S |
| Molecular Weight (g/mol) | 136.13 |
| MDL Number | MFCD00010912 |
| InChI Key | OSGAYBCDTDRGGQ-UHFFFAOYSA-L |
| PubChem CID | 24497 |
| ChEBI | CHEBI:31346 |
| IUPAC Name | calcium sulfate |
| SMILES | [Ca++].[O-]S([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
2002762
|
Avantor J.T.Baker
L05607 |
454 g |
Case of 12 Each for $7,151.00
|
|
|||||
|
02-002-761
|
Avantor
L05602 |
2.3 kg |
Case of 4 Each for $7,430.22
|
|
|||||
Chemical Identifiers
| 7778-18-9 | |
| 136.13 | |
| OSGAYBCDTDRGGQ-UHFFFAOYSA-L | |
| CHEBI:31346 | |
| [Ca++].[O-]S([O-])(=O)=O |
| CaO4S | |
| MFCD00010912 | |
| 24497 | |
| calcium sulfate |
Specifications
| 7778-18-9 | |
| CaO4S | |
| 8 Mesh | |
| [Ca++].[O-]S([O-])(=O)=O | |
| 136.13 | |
| CHEBI:31346 | |
| DRIERITE™, Non-Indicating |
| 454 g | |
| MFCD00010912 | |
| OSGAYBCDTDRGGQ-UHFFFAOYSA-L | |
| calcium sulfate | |
| 24497 | |
| 1L = 2.96kg |