missing translation for 'onlineSavingsMsg'
Learn More

DRIERITE™, Non-Indicating, 8 Mesh, J.T. Baker™

Supplier:  Avantor J.T.Baker L05602

 View more versions of this product

Catalog No. 02-002-761


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

DRIERITE™
7778-18-9
CaO4S
8 Mesh
[Ca++].[O-]S([O-])(=O)=O
136.13
CHEBI:31346
Non-Indicating
2.3 kg
MFCD00010912
OSGAYBCDTDRGGQ-UHFFFAOYSA-L
calcium sulfate
24497
1L = 2.96kg

Chemical Identifiers

7778-18-9
136.13
OSGAYBCDTDRGGQ-UHFFFAOYSA-L
CHEBI:31346
[Ca++].[O-]S([O-])(=O)=O
CaO4S
MFCD00010912
24497
calcium sulfate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions