missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 5 N Aqueous Solution, Spectrum™ Chemical

Catalog No. 18606558
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
4 x 4 L
6 x 1 L
55 gal.
Packaging:
Poly Bottle
Poly Drum
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606558 6 x 1 L Poly Bottle
18606552 1 L Poly Bottle
18606553 4 L Poly Bottle
18606554 4 x 4 L Poly Bottle
18606555 20 L Poly Bottle
18606557 55 gal. Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18606558 Supplier Spectrum Chemical Mfg Cor Supplier No. S3901LTCS6

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

S-390, 1310-73-2

Spectrum™ Chemical Sodium Hydroxide, 5 N Aqueous Solution is a standard solution of Caustic soda ideally suited for laboratory titrimetric techniques. You can rely on the quality of LabChem analytical test solutions for lot-to-lot reproducibility.

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 40.00
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
IUPAC Name sodium hydroxide
SMILES [OH-].[Na+]

Specifications

CAS Max % 0.17, 0.83
Concentration 5.0 N ± 0.2 N
Quantity 6 x 1 L
Grade Reagent
Packaging Poly Bottle
Chemical Name or Material Sodium Hydroxide Solution