missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DreamTaq™ Hot Start DNA Polymerase

Thermo Scientific DreamTaq Hot Start DNA Polymerase is an enhanced hot start Taq DNA polymerase that enables higher PCR specificity, sensitivity, and yields compared to conventional hot start Taq DNA polymerases.

Supplier:  Thermo Scientific™ EP1711

 View more versions of this product

Catalog No. FEREP1711


Only null left

Description

Description

Thermo Scientific DreamTaq Hot Start DNA Polymerase, available in colorless and green formats, is an enhanced hot start Taq DNA polymerase that enables higher PCR specificity, sensitivity, and yields compared to conventional hot start Taq DNA polymerases.

DreamTaq Hot Start DNA Polymerase employs antibody-based inhibition of DNA polymerase activity at ambient temperatures to prevent the amplification of non-specific products prior to the amplification step. With DreamTaq Hot Start DNA Polymerase, reactions can be set up at room temperature using the same protocol and cycling conditions as conventional Taq DNA polymerases. The enzyme is supplied with an optimized 10X DreamTaq Buffer containing magnesium chloride, which eliminates the need for extensive optimization of reaction conditions. DreamTaq Hot Start DNA Polymerase is also available with pre-added density reagent and electrophoresis tracking dyes, as well as in master mix format.

The 10X DreamTaq Green Buffer also includes a density reagent and two tracking dyes for convenient direct loading of PCR products on a gel.

DreamTaq Hot Start DNA Polymerase is formulated to enhance productivity through:

  • Better reaction outcomes
    • Higher yield of target amplicons from low template amounts
    • Increased reaction specificity due to reliable hot-start technology
    • Wider range of amplicon lengths with routine amplification of genomic DNA fragments up to 6 kb
  • Enhanced convenience
    • Room temperature reaction setup
    • Minimized optimization of Mg2+ concentration and of primer annealing temperatures due to optimized reaction buffer
    • Direct loading of PCR products on a gel with DreamTaq Green Buffer

Applications

  • Generation of PCR products for TA cloning
  • Routine PCR
  • Colony PCR
  • Genotyping
  • RT-PCR

Recommended Storage

Store at -20C.

Specifications

Chemical Identifiers

5 U/μL
DreamTaq DNA Polymerase
Hot-start PCR
3'-A
Low
Standard
Built-In Hot Start
Separate Components

Specifications

5 U/μL
Low
Standard
Dry Ice
Built-In Hot Start
Separate Components
• DreamTaq Hot Start DNA Polymerase (5 U/μL), 40 μL
• 10X DreamTaq Green Buffer (with 20 mM MgCl2), 1.25 mL

Store at -20°C.
DreamTaq DNA Polymerase
200 U
Hot-start PCR
3'-A
Green
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions