Learn More
Dräger™ Short-Term Detector Tubes: Water Vapor
Detector tubes
$186.80 - $274.72
Specifications
Components Measured Water Vapor
Quantity 10
View More Specs
Products 6
Catalog Number Mfr. No. Measurement Range Tube Type Response Time Price Quantity & Availability  
Catalog Number Mfr. No. Measurement Range Tube Type Response Time Price Quantity & Availability  
29663376
View Documents Promotion Details
Dräger™
6728531
5 to 200 Water vapor 5/a-p 25 min.
Pack of 10 Each for $205.10
Only null left
18556N
View Documents Promotion Details
Dräger™
8103031
3 to 60 Water vapor 3/a Not Applicable
Pack of 10 Each for $270.74
Only null left
17985317
View Documents Promotion Details
Dräger™
8101321
0.05 to 1 Water vapor 0.1/a 1.5 min.
Pack of 10 Each for $240.91
Only null left
18556Q
View Documents Promotion Details
Dräger™
8101781
20 to 40mg/L Water vapor 1/b 20/40 sec.
Pack of 10 Each for $240.91
Only null left
19046633
View Documents Promotion Details
Dräger™
8103061
20 to 1500 Water vapor 20/a-P 10 to 5 min.
Pack of 10 Each for $186.80
Only null left
17985319
View Documents Promotion Details
Dräger™
CH23401
1 to 40 Water vapor 0.1 2 min.
Pack of 10 Each for $274.72
Only null left
Description
Description
Detector tubes
Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.
Note: Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.
Order Info
Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ Customer Service for more information.
Specifications
Specifications
Water Vapor
10