Learn More
Dräger™ Short-Term Detector Tubes: Water Vapor
Detector tubes
Supplier:  Dräger™ 8101321
 View more versions of this product
Catalog No. 17985317
Only null left
Description
Description
Detector tubes
Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.
Note: Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.
Order Info
Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ Customer Service for more information.
Specifications
Chemical Identifiers
Water Vapor
10
Specifications
Water Vapor
0.05 to 1
10
32°F to 86°F
Water vapor 0.1/a
Not Applicable
3
1.5 min.
0°C to 30°C
Product Suggestions
Product Suggestions