Learn More
Dräger™ CMS Chips: Benzene
Suitable for 10 measurements of one gas
Supplier: Dräger™ 6406030
Description
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.Chemical Identifiers
| 700 to 1100 hpa | |
| Benzene CMS Chip | |
| 32°F to 104°F | |
| 0°C to 40°C |
Specifications
| 700 to 1100 hpa | |
| 1 to 30mg/L | |
| 35 to 300 sec. | |
| 0°C to 40°C |
| Benzene CMS Chip | |
| 0.2 to 10 | |
| 32°F to 104°F |