Learn More
Dräger™ CMS Chips: Hydrogen Sulfide
Suitable for 10 measurements of one gas.
Supplier:  Dräger™ 6406150
 View more versions of this product
Catalog No. 189991927
Only null left
Description
Description
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Chemical Identifiers
700 to 1100 hpa
Hydrogen Sulphide CMS Chip
32°F to 104°F
0°C to 40°C
Specifications
700 to 1100 hpa
1 to 40mg/L
30 to 240 sec.
0°C to 40°C
Hydrogen Sulphide CMS Chip
20 to 500
32°F to 104°F
Product Suggestions
Product Suggestions