missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DPD Indicator Solution, for Residual Chlorine Analysis, Ricca Chemical
For Analysis of Residual Chlorine Analysis
$80.50 - $1081.50
Specifications
| Chemical Name or Material | DPD Indicator Solution |
|---|---|
| CAS | 7732-18-5 , 7664-93-9 , 6283-63-2 , 6381-92-6 |
| CAS Min % | 98.0,0.33,0.1,0.02 |
| CAS Max % | 100.0,0.34,0.1,0.02 |
| Grade | Laboratory |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
R2655000100
|
Ricca Chemical Company
R2655000100C |
100 mL |
Each for $80.50
|
|
|||||
|
2655-16
|
Ricca Chemical Company
265516 |
500 mL |
Each for $253.57
|
|
|||||
|
2655-32
|
Ricca Chemical Company
265532 |
1 L |
Each for $361.17
|
|
|||||
|
26551
|
Ricca Chemical Company
26551 |
4 L |
Each for $1,081.50
|
|
|||||