missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dorsomorphin 97.0+%, TCI America™
$393.32 - $1181.81
Chemical Identifiers
| CAS | 866405-64-3 |
|---|---|
| Molecular Formula | C24H25N5O |
| Molecular Weight (g/mol) | 399.50 |
| MDL Number | MFCD08705402 |
| InChI Key | XHBVYDAKJHETMP-UHFFFAOYSA-N |
| Synonym | 6-[4-(2-Piperidin-1-ylethoxy)phenyl]-3-pyridin-4-ylpyrazolo[1,5-a]pyrimidine |
| PubChem CID | 11524144 |
| ChEBI | CHEBI:78510 |
| IUPAC Name | 4-(6-{4-[2-(piperidin-1-yl)ethoxy]phenyl}pyrazolo[1,5-a]pyrimidin-3-yl)pyridine |
| SMILES | C(CN1CCCCC1)OC1=CC=C(C=C1)C1=CN2N=CC(=C2N=C1)C1=CC=NC=C1 |
Chemical Identifiers
| 866405-64-3 | |
| 399.50 | |
| XHBVYDAKJHETMP-UHFFFAOYSA-N | |
| 11524144 | |
| 4-(6-{4-[2-(piperidin-1-yl)ethoxy]phenyl}pyrazolo[1,5-a]pyrimidin-3-yl)pyridine |
| C24H25N5O | |
| MFCD08705402 | |
| 6-[4-(2-Piperidin-1-ylethoxy)phenyl]-3-pyridin-4-ylpyrazolo[1,5-a]pyrimidine | |
| CHEBI:78510 | |
| C(CN1CCCCC1)OC1=CC=C(C=C1)C1=CN2N=CC(=C2N=C1)C1=CC=NC=C1 |
Specifications
| 866405-64-3 | |
| MFCD08705402 | |
| 6-[4-(2-Piperidin-1-ylethoxy)phenyl]-3-pyridin-4-ylpyrazolo[1,5-a]pyrimidine | |
| C(CN1CCCCC1)OC1=CC=C(C=C1)C1=CN2N=CC(=C2N=C1)C1=CC=NC=C1 | |
| 399.50 | |
| CHEBI:78510 | |
| ≥97.0% (HPLC) | |
| Dorsomorphin |
| C24H25N5O | |
| 10 mg | |
| XHBVYDAKJHETMP-UHFFFAOYSA-N | |
| 4-(6-{4-[2-(piperidin-1-yl)ethoxy]phenyl}pyrazolo[1,5-a]pyrimidin-3-yl)pyridine | |
| 11524144 | |
| 399.50 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator