missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dorsomorphin 97.0+%, TCI America™
Supplier: TCI America D539450MG
Specifications
| Dorsomorphin | |
| C24H25N5O | |
| 50 mg | |
| XHBVYDAKJHETMP-UHFFFAOYSA-N | |
| 4-(6-{4-[2-(piperidin-1-yl)ethoxy]phenyl}pyrazolo[1,5-a]pyrimidin-3-yl)pyridine | |
| 11524144 | |
| 399.50 | |
| Crystalline Powder |
| 866405-64-3 | |
| MFCD08705402 | |
| 6-[4-(2-Piperidin-1-ylethoxy)phenyl]-3-pyridin-4-ylpyrazolo[1,5-a]pyrimidine | |
| C(CN1CCCCC1)OC1=CC=C(C=C1)C1=CN2N=CC(=C2N=C1)C1=CC=NC=C1 | |
| 399.50 | |
| CHEBI:78510 | |
| ≥97.0% (HPLC) |
Chemical Identifiers
| 866405-64-3 | |
| 399.50 | |
| XHBVYDAKJHETMP-UHFFFAOYSA-N | |
| 11524144 | |
| 4-(6-{4-[2-(piperidin-1-yl)ethoxy]phenyl}pyrazolo[1,5-a]pyrimidin-3-yl)pyridine |
| C24H25N5O | |
| MFCD08705402 | |
| 6-[4-(2-Piperidin-1-ylethoxy)phenyl]-3-pyridin-4-ylpyrazolo[1,5-a]pyrimidine | |
| CHEBI:78510 | |
| C(CN1CCCCC1)OC1=CC=C(C=C1)C1=CN2N=CC(=C2N=C1)C1=CC=NC=C1 |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator