Learn More
MilliporeSigma™ Supelco™ domnick hunter™ NITROX™ UHP Hydrogen Generator
A safe and economic alternative to high pressure gas cylinders
Supplier: MilliporeSigma™ Supelco™ 27750U
Description
- A safe and economic alternative to high pressure gas cylinders
- Delivers a continuous stream of 99.999% pure hydrogen
- Ideal for carrier gas and fuel gas applications
- Suitable for use with any laboratory instrument requiring a supply of ultra high purity hydrogen
- Utilizes a proprietary ion exchange membrane to produce ultra-high-purity hydrogen from laboratory grade deionized water, totally eliminating the need for a caustic solution
- Hydrogen is generated only on demand, at low pressure, and minimal volume is stored within the unit
- The unique digital display gives system status and diagnostic capabilities not available on any other competitive hydrogen generator
- Designed for total safety and highly reliable operation
Chemical Identifiers
| Any Laboratory Instrument Requiring a Supply of Ultra High Purity Hydrogen | |
| Carrier Gas and Fuel Gas Applications | |
| 99.999% Pure Hydrogen | |
| Hydrogen Gas Generator |
Specifications
| 0-500 cc/min | |
| 60 H | |
| 99.999% Pure Hydrogen |
| Any Laboratory Instrument Requiring a Supply of Ultra High Purity Hydrogen | |
| Carrier Gas and Fuel Gas Applications | |
| Hydrogen Gas Generator |
NITROX is a trademark of domnick hunter group PLC